Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SCUBE3 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP241186
Description
SCUBE3 Polyclonal specifically detects SCUBE3 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
SCUBE3 | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
SCUBE3 | |
Antibody was raised against a 15 amino acid synthetic peptide near the center of human SCUBE3. The immunogen is located within amino acids 210 - 260 of SCUBE3. Amino Acid Squence: HIKFVLHTDGKT | |
Affinity Purified | |
RUO | |
222663 | |
Human, Mouse | |
IgG |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
1 mg/mL | |
Western Blot 1-2 ug/ml, ELISA, Immunocytochemistry/Immunofluorescence 20 ug/mL | |
CEGF3, CUB and EGF containing protein 3, CUB domain and EGF-like repeat containing 3, DKFZp686B09105, DKFZp686B1223, DKFZp686D20108, FLJ34743, signal peptide, CUB and EGF-like domain containing protein 3, signal peptide, CUB and EGF-like domain-containing protein 3, signal peptide, CUB domain, EGF-like 3 | |
Rabbit | |
109 kDa | |
0.1 mg | |
Primary | |
At least two isoforms of SCUBE3 are known to exist. SCUBE3 antibody is predicted to not cross-react with other SCUBE members | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction