Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SCUBE3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA534427
Description
A suggested positive control is mouse kidney tissue lysate. PA5-34427 can be used with blocking peptide PEP-1469.
SCUBE3 is a member of a family of secreted glycoproteins that contain N-terminal EGF-like repeats and C-terminal cysteine-rich motifs and CUB domain and is highly expressed in primary osteoblasts and bones, and to a lesser extent in heart. Other studies have shown that overexpression of SCUBE3 in mice induced cardiac hypertrophy, suggesting that it may also play a role in the regulation of cardiac growth. SCUBE3 has been shown to be an endogenous TGF-Beta receptor ligand and is thought to promote lung cancer cell mobility and invasiveness. In lung cancer cells, the secreted SCUBE3 protein was cleaved by MMP2 and MMP9, allowing the activation of the TGF-Beta receptor, the increase of Smad2/3 transcriptional activity and the upregulation of expression of proteins such as TGF-Beta1, VEGF, Snail, and Slug.
Specifications
SCUBE3 | |
Polyclonal | |
Unconjugated | |
SCUBE3 | |
CEGF3; CUB domain and EGF-like repeat containing 3; D030038I21Rik; SCUBE3; signal peptide; signal peptide, CUB and EGF-like domain containing protein 3; signal peptide, CUB and EGF-like domain-containing protein 3; signal peptide, CUB domain and EGF like domain containing 3; signal peptide, CUB domain, EGF-like 3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
222663, 268935 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q66PY1, Q8IX30 | |
SCUBE3 | |
A 15 amino acid synthetic peptide near the center of human SCUBE3. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction