Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCUBE3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534558
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534558 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534558 Supplier Thermo Scientific Supplier No. PA534558
Add to Cart
Add to Cart

SCUBE3 Polyclonal Antibody

SCUBE3 Polyclonal Antibody for IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen SCUBE3
Applications Immunohistochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene SCUBE3
Gene Accession No. Q8IX30
Gene Alias CEGF3; CUB domain and EGF-like repeat containing 3; D030038I21Rik; SCUBE3; signal peptide; signal peptide, CUB and EGF-like domain containing protein 3; signal peptide, CUB and EGF-like domain-containing protein 3; signal peptide, CUB domain and EGF like domain containing 3; signal peptide, CUB domain, EGF-like 3
Gene Symbols SCUBE3
Host Species Rabbit
Immunogen A 15 amino acid peptide near the center of human SCUBE3.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 222663
Target Species Human
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.