Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SCX Antibody (PCRP-SCXA-1D2), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314175PCP
This item is not returnable.
View return policy
Description
SCX Monoclonal antibody specifically detects SCX in Human samples. It is validated for Flow CytometrySpecifications
SCX | |
Monoclonal | |
PerCP | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
642658 | |
Store at 4°C in the dark. | |
Purified |
Flow Cytometry | |
PCRP-SCXA-1D2 | |
Flow Cytometry | |
BHLHA41, BHLHa48, Class A Basic Helix-Loop-Helix Protein 41, Class A Basic Helix-Loop-Helix Protein 48, Class II BHLH Protein Scleraxis, Scleraxis Basic Helix-Loop-Helix Transcription Factor, Scleraxis BHLH Transcription Factor, Scleraxis Homolog (Mouse), Scleraxis Homolog A, Scleraxis Homolog A (Mouse), Scleraxis Homolog B, Scleraxis Homolog B (Mouse), SCXA, SCXB | |
Recombinant full-length human SCX protein (Uniprot: Q7RTU7) | |
0.1 mL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction