Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SCX Mouse anti-Human, Clone: PCRP-SCXA-1D2, Novus Biologicals™

Mouse Monoclonal Antibody
$240.00 - $492.50
Specifications
| Antigen | SCX |
|---|---|
| Clone | PCRP-SCXA-1D2 |
| Concentration | 0.2 mg/ml |
| Dilution | Flow Cytometry 1-2 ug/million cells, Immunofluorescence 1-2 ug/ml |
| Applications | Flow Cytometry, Immunofluorescence |
Description
SCX Monoclonal antibody specifically detects SCX in Human samples. It is validated for Flow Cytometry, ImmunofluorescenceSpecifications
| SCX | |
| 0.2 mg/ml | |
| Flow Cytometry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q7RTU7 | |
| 642658 | |
| IgG2a | |
| Protein A or G purified |
| PCRP-SCXA-1D2 | |
| Flow Cytometry 1-2 ug/million cells, Immunofluorescence 1-2 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| BHLHA41, BHLHa48, Class A Basic Helix-Loop-Helix Protein 41, Class A Basic Helix-Loop-Helix Protein 48, Class II BHLH Protein Scleraxis, Scleraxis Basic Helix-Loop-Helix Transcription Factor, Scleraxis BHLH Transcription Factor, Scleraxis Homolog (Mouse), Scleraxis Homolog A, Scleraxis Homolog A (Mouse), Scleraxis Homolog B, Scleraxis Homolog B (Mouse), SCXA, SCXB | |
| Recombinant full-length human SCX protein (Uniprot: Q7RTU7) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title