Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SCXA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115874
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115874 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115874 Supplier Invitrogen™ Supplier No. PA5115874
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SCXA.

Enables DNA-binding transcription activator activity, RNA polymerase II-specific and bHLH transcription factor binding activity. Contributes to E-box binding activity. Involved in positive regulation of transcription by RNA polymerase II. Located in nucleus. Part of transcription regulator complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCXA
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene SCX
Gene Accession No. Q64124, Q7RTU7
Gene Alias basic helix-loop-helix transcription factor scleraxis; BB114693; Bhlha41; BHLHA48; class A basic helix-loop-helix protein 41; Class A basic helix-loop-helix protein 48; class II bHLH protein scleraxis; Scl; scleraxis; scleraxis basic helix-loop-helix transcription factor; scleraxis bHLH transcription factor; scleraxis homolog A; scleraxis homolog B; SCX; SCXA; SCXB
Gene Symbols SCX
Host Species Rabbit
Immunogen A synthesized peptide derived from human SCX(Accession Q7RTU7), corresponding to amino acid residues R28-H78.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20289, 642658, 680712
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.