Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SCXA Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA523943
Description
This antibody is predicted to react with chicken based on sequence homology.
Enables DNA-binding transcription activator activity, RNA polymerase II-specific and bHLH transcription factor binding activity. Contributes to E-box binding activity. Involved in positive regulation of transcription by RNA polymerase II. Located in nucleus. Part of transcription regulator complex.
Specifications
| SCXA | |
| Polyclonal | |
| Unconjugated | |
| SCX | |
| basic helix-loop-helix transcription factor scleraxis; BB114693; Bhlha41; BHLHA48; class A basic helix-loop-helix protein 41; Class A basic helix-loop-helix protein 48; class II bHLH protein scleraxis; Scl; scleraxis; scleraxis basic helix-loop-helix transcription factor; scleraxis bHLH transcription factor; scleraxis homolog A; scleraxis homolog B; SCX; SCXA; SCXB | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 20289, 642658 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q64124, Q7RTU7 | |
| SCX | |
| KLH conjugated synthetic peptide between 172-201 amino acids from the C-terminal region of human SCXA. | |
| 200 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction