Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SCYL1 Antibody, Alexa Fluor™ 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297454AF405
Description
SCYL1 Polyclonal specifically detects SCYL1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
SCYL1 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
Coated vesicle-associated kinase of 90 kDa, CVAK90, GKLPTelomerase transcriptional element-interacting factor, likely ortholog of mouse N-terminal kinase-like protein, MGC78454, NKTL, N-terminal kinase-like protein, NTKLN-terminal kinase-like, P105, SCY1-like 1 (S. cerevisiae), TAPKTeratoma-associated tyrosine kinase, TEIFSCY1-like protein 1, Telomerase regulation-associated protein, telomerase transcriptional elements-interacting factor, TRAPHT019 | |
Produced in rabbits immunized with E. coli-derived Human SCYL1 fragment. | |
0.1 mL | |
Protein Kinase | |
57410 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 405 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction