Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SCYL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA553267
Description
Immunogen sequence: AFPEDFCRHK VLPQLLTAFE FGNAGAVVLT PLFKVGKFLS AEEYQQKIIP VVVKMFSSTD RAMRIRLLQQ MEQFIQYLDE PTVNTQIFPH VVHGFLDTNP AIREQTVKSM LLLAPKLNEA NLNVELMK Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 95%.
With approximately 135 million base pairs and 1,400 genes, chromosome 11 makes up around 4% of human genomic DNA and is considered a gene and disease association dense chromosome. The chromosome 11 encoded Atm gene is important for regulation of cell cycle arrest and apoptosis following double strand DNA breaks. Atm mutation leads to the disorder known as ataxia-telangiectasia. The blood disorders Sickle cell anemia and β thalassemia are caused by HBB gene mutations. Wilms' tumors, WAGR syndrome and Denys-Drash syndrome are associated with mutations of the WT1 gene. Jervell and Lange-Nielsen syndrome, Jacobsen syndrome, Niemann-Pick disease, hereditary angioedema and Smith-Lemli-Opitz syndrome are also associated with defects in chromosome 11.
Specifications
| SCYL1 | |
| Polyclonal | |
| Unconjugated | |
| Scyl1 | |
| 105 kDa kinase-like protein; 2810011O19Rik; C85140; Coated vesicle-associated kinase of 90 kDa; CVAK90; GKLP; HT019; likely ortholog of mouse N-terminal kinase-like protein; mdf; mfd; Mitosis-associated kinase-like protein NTKL; NKTL; N-terminal kinase-like protein; NTKL; P105; SCAR21; SCY1 like pseudokinase 1; SCY1-like 1 (S. cerevisiae); SCY1-like 1 splice variant 13; SCY1-like 1 splice variant 15; SCY1-like protein 1; SCY1-like, kinase-like 1; Scyl1; TAPK; TEIF; Telomerase regulation-associated protein; Telomerase transcriptional element-interacting factor; telomerase transcriptional elements-interacting factor; teratoma-associated tyrosine kinase; TRAP; ubiquitous protein kinase-like (105 kDa) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 57410 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96KG9 | |
| Scyl1 | |
| Recombinant protein corresponding to Human SCYL1. Recombinant protein control fragment (Product #RP-91451). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction