Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCYL1 Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. ABS149425UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS1494-25UG 25 μg
ABS1494-100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ABS1494-25UG Supplier MilliporeSigma Supplier No. ABS149425UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-SCYL1 is a rabbit polyclonal antibody that detects SCY1-Like Protein 1 and has been tested for use in Immunocytochemistry and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCYL1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols SCYL1,CVAK90,GKLP,NTKL,TAPK,TEIF,TRAP,HT01
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to 16 amino acids from the C-terminal region of human SCY1-Like Protein 1.
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_001041683.1
Test Specificity This rabbit polyclonal antibody detects SCY1-Like Protein 1 (SCYL1) in human cells. It targets an epitope within 16 amino acids from the C-terminal region. The immunogen is conserved between isoforms 1, 2, and 3.
Target Species Human, Mouse
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.