Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SCYL1BP1 Antibody (OTI2H6), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP203929
Description
SCYL1BP1 Monoclonal specifically detects SCYL1BP1 in Human samples. It is validated for Western Blot.Specifications
SCYL1BP1 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:2000 | |
GORAB | |
Human recombinant protein fragment corresponding to amino acids 35-394 of human GORAB (NP_689494) produced in E.coli. | |
Affinity Purified | |
RUO | |
92344 | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot | |
OTI2H6 | |
Unconjugated | |
FLJ11752, golgin, RAB6-interacting, GONTKL-binding protein 1, hNTKL-BP1, MGC51263, MGC70512, N-terminal kinase-like-binding protein 1, NTKL-BP1, NTKLBP1SCY1-like 1-binding protein 1, RAB6-interacting golgin, SCY1-like 1 binding protein 1, SCYL1-BP1, SCYL1BP1SCYL1-binding protein 1 | |
Mouse | |
44.8 kDa | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction