Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SCYL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554317
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554317 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554317 Supplier Invitrogen™ Supplier No. PA554317
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LNVTPTVRPD ADQMTKIPFF DDVGAVTLQY FDTLFQRDNL QKSQFFKGLP KVLPKLPKRV IVQRILPCLT SEFVNPDMVP FVLPNVLLIA EECTKEEYVK LILPELGPVF KQQEPIQILL IFLQKMDLLL TKTPPDEIKN S Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.

SCYL2 is a component of AP2-containing clathrin coated structures at the plasma membrane or of endocytic coated vesicles. SCYL2 is a probable serine/threonine-protein kinase that phosphorylates, in vitro, the beta2-subunit of the plasma membrane adapter complex AP2 and other proteins in presence of poly-L-lysine. SCYL2 may also regulate clathrin-dependent trafficking between the TGN and/or the endosomal system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCYL2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Scyl2
Gene Accession No. Q6P3W7
Gene Alias AU015688; AW536445; BC030932; Coated vesicle-associated kinase of 104 kDa; CVAK104; D10Ertd802e; KIAA1360; RGD1305665; SCY1 like pseudokinase 2; SCY1-like 2; SCY1-like 2 (S. cerevisiae); SCY1-like protein 2; SCY1-like, kinase-like 2; SCYL2
Gene Symbols Scyl2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SCYL2. Recombinant protein control fragment (Product #RP-91441).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55681
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.