Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SCYL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589051
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589051 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589051 Supplier Invitrogen™ Supplier No. PA589051
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MESMLNKLKS TVTKVTADVT SAVMGNPVTR EFDVGRHIAS GGNGLAWKIF NGTKKSTKQE VAVFVFDKKL IDKYQKFEKD QIIDSLKRGV QQLTRLRHPR LLTVQHPLEE SRDCLAFCTE PVFASLANVL GNWENLPSPI SPDIKDYKLY DVETKYGLLQ VSEGLSFLHS SVKMVHGNIT PENIILNKSG AWKIMGFDFC VSSTNPSEQE PKFPCKEWDP NLPSLCLPNP EYLAPEYILS VSCETASDMY SLGTVMYAVF NKGKPIFEVN KQDIYKSFSR QLDQLSRLGS SSLTNIPEEV; Positive Samples: Mouse brain, Mouse heart, Mouse kidney, Rat brain, Rat heart; Cellular Location: Cytoplasm, Cytoplasmic vesicle, Endosome membrane, Golgi apparatus, clathrin-coated vesicle, perinuclear region, trans-Golgi network membrane.

SCYL2 is a component of AP2-containing clathrin coated structures at the plasma membrane or of endocytic coated vesicles. SCYL2 is a probable serine/threonine-protein kinase that phosphorylates, in vitro, the beta2-subunit of the plasma membrane adapter complex AP2 and other proteins in presence of poly-L-lysine. SCYL2 may also regulate clathrin-dependent trafficking between the TGN and/or the endosomal system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCYL2
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.26 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Scyl2
Gene Accession No. Q6P3W7, Q8CFE4
Gene Alias AU015688; AW536445; BC030932; Coated vesicle-associated kinase of 104 kDa; CVAK104; D10Ertd802e; KIAA1360; RGD1305665; SCY1 like pseudokinase 2; SCY1-like 2; SCY1-like 2 (S. cerevisiae); SCY1-like protein 2; SCY1-like, kinase-like 2; SCYL2
Gene Symbols Scyl2
Host Species Rabbit
Immunogen Recombinant protein (or fragment)
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 213326, 314717, 55681
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.