Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCYL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589052
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589052 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589052 Supplier Thermo Scientific Supplier No. PA589052
Add to Cart
Add to Cart

SCYL2 Polyclonal Antibody

SCYL2 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen SCYL2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.42 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Scyl2
Gene Accession No. Q6P3W7
Gene Alias AU015688; AW536445; BC030932; Coated vesicle-associated kinase of 104 kDa; CVAK104; D10Ertd802e; KIAA1360; RGD1305665; SCY1 like pseudokinase 2; SCY1-like 2; SCY1-like 2 (S. cerevisiae); SCY1-like protein 2; SCY1-like, kinase-like 2; SCYL2
Gene Symbols Scyl2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human SCYL2 (NP_0604583).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 314717, 55681
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.