Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SCYL3 Monoclonal Antibody (OTI1A5)
GREENER_CHOICE

Catalog No. PIMA524931
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524931 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524931 Supplier Invitrogen™ Supplier No. MA524931
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SCYL3 Monoclonal Antibody for Western Blot, IHC (P)

May play a role in regulating cell adhesion/migration complexes in migrating cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCYL3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1A5
Concentration 0.85 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SCYL3
Gene Accession No. Q8IZE3
Gene Alias 1200016D23Rik; 6030457O16; AW214499; ezrin-binding partner PACE-1; ezrin-binding partner PACE-1 (PACE-1); ezrin-binding protein PACE-1; Pace1; PACE-1; protein-associating with the carboxyl-terminal domain of ezrin; RP1-97P20.2; SCY1 like pseudokinase 3; SCY1-like 3; SCY1-like 3 (S. cerevisiae); SCY1-like protein 3; SCY1-like, kinase-like 3; Scyl3
Gene Symbols SCYL3
Host Species Mouse
Immunogen Full-length protein expressed in 293T cell transfected with human SCYL3 expression vector.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57147
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.