Learn More
Invitrogen™ SCYL3 Polyclonal Antibody

Description
SCYL3 Polyclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | SCYL3 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | SCYL3 |
| Gene Accession No. | Q9DBQ7 |
| Gene Alias | 1200016D23Rik; 6030457O16; AW214499; ezrin-binding partner PACE-1; ezrin-binding partner PACE-1 (PACE-1); ezrin-binding protein PACE-1; Pace1; PACE-1; protein-associating with the carboxyl-terminal domain of ezrin; RP1-97P20.2; SCY1 like pseudokinase 3; SCY1-like 3; SCY1-like 3 (S. cerevisiae); SCY1-like protein 3; SCY1-like, kinase-like 3; Scyl3 |
| Gene Symbols | SCYL3 |
| Host Species | Rabbit |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.