Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCYL3 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501727001 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-7001 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-7001 Supplier Proteintech Group Inc Supplier No. 112491AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

May play a role in regulating cell adhesion/migration complexes in migrating cells.

Specifications

Antigen SCYL3
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.19 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene SCYL3
Gene Accession No. Q8IZE3
Gene Alias Ezrin binding protein PACE 1, PACE 1, PACE1, RP1 97P20.2, SCY1 like 3 (S. cerevisiae), SCY1 like protein 3, SCYL3
Gene Symbols SCYL3
Host Species Rabbit
Immunogen SCYL3 Fusion Protein Ag1769
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57147
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.