Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ SD 1008
GREENER_CHOICESDP

Catalog No. 303550 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30-355-0 50 mg
30-351-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 30-355-0 Supplier R&D Systems™ Supplier No. 3035/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. JAK2/STAT3 signaling pathway inhibitor

JAK2/STAT3 signaling pathway inhibitor. Inhibits activation of STAT3, JAK2 and Src. Induces apoptosis in cell lines expressing constitutively active tyrosine-phosphorylated STAT3.

Chemical Identifiers

CAS 960201-81-4
Molecular Formula C18H19NO5

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 329.35
Purity >99%
Molecular Weight (g/mol) 329.352
Chemical Name or Material SD 1008
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.