Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SDC3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | SDC3 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
SDC3 Polyclonal specifically detects SDC3 in Mouse samples. It is validated for Western Blot.Specifications
| SDC3 | |
| Polyclonal | |
| Rabbit | |
| Q64519 | |
| 9672 | |
| Synthetic peptides corresponding to the N terminal of Sdc3. Immunizing peptide sequence GLLLPPLLLLLLAGRAAGAQRWRNENFERPVDLEGSGDDDSFPDDELDDL. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| KIAA0468, N-syndecan, SDCN, SYND3syndecan proteoglycan 3, syndecan 3, syndecan 3 (N-syndecan), syndecan neural type, syndecan-3 | |
| SDC3 | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title