Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SDC3 Antibody (SR2066), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32256650UL
Description
SDC3 Monoclonal antibody specifically detects SDC3 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
SDC3 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
9672 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Western Blot | |
SR2066 | |
Western Blot 1:500-1:2000 | |
KIAA0468, N-syndecan, SDCN, SYND3syndecan proteoglycan 3, syndecan 3, syndecan 3 (N-syndecan), syndecan neural type, syndecan-3 | |
A synthesized peptide derived from human SDC3 (Uniprot #: O75056) | |
50 μg | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction