Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SDCCAG10 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$646.00
Specifications
Antigen | SDCCAG10 |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
SDCCAG10 Polyclonal specifically detects SDCCAG10 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
SDCCAG10 | |
Polyclonal | |
Rabbit | |
Human, Mouse, Rat | |
Antigen NY-CO-10, CWC27 spliceosome-associated protein homolog (S. cerevisiae), EC 5.2.1.8, NY-CO-10, peptidyl-prolyl cis-trans isomerase SDCCAG10, PPIase CWC27, PPIase SDCCAG10, SDCCAG10, Serologically defined colon cancer antigen 10peptidyl-prolyl cis-trans isomerase CWC27 homolog | |
CWC27 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
10283 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:REDQTLALLNQFKSKLTQAIAETPENDIPETEVEDDEGWMSHVLQFEDKSRKVKDASMQDSDTFEIYDPRNPVNKRRREESK | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title