Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SDCCAG10 Antibody (OTI4C8), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274031AF594
Description
SDCCAG10 Monoclonal specifically detects SDCCAG10 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
SDCCAG10 | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
CWC27 | |
Recombinant protein fragment corresponding to amino acids 145-472 of human SDCCAG10 (NP_005860) produced in E.coli. | |
0.1 mL | |
10283.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI4C8 | |
Western Blot, Immunohistochemistry | |
Antigen NY-CO-10, CWC27 spliceosome-associated protein homolog (S. cerevisiae), EC 5.2.1.8, NY-CO-10, peptidyl-prolyl cis-trans isomerase SDCCAG10, PPIase CWC27, PPIase SDCCAG10, SDCCAG10, Serologically defined colon cancer antigen 10peptidyl-prolyl cis-trans isomerase CWC27 homolog | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction