Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SDCCAG3 Isoform 1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115876
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115876 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115876 Supplier Invitrogen™ Supplier No. PA5115876
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SDCCAG3.

SDCCAG3, also named as NY-CO-3, is identified as serologically defined colon cancer antigen-3. It is an endosomal protein which is involved in the regulation of cytokinesis. SDCCAG3 is involved in modulation of TNF response. This gene has four isoforms with MW 40-50 kDa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SDCCAG3 Isoform 1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene ENTR1
Gene Accession No. Q96C92
Gene Alias Antigen NY-CO-3; C330016H24Rik; C630038K21Rik; Endosome-associated-trafficking regulator 1; Entr1; NY-CO-3; RP11-413M3.3; Sdccag3; Serologically defined colon cancer antigen 3; serologically defined colon cancer antigen 3 homolog
Gene Symbols ENTR1
Host Species Rabbit
Immunogen A synthesized peptide derived from human SDCCAG3(Accession Q96C92), corresponding to amino acid residues A57-N107.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10807, 306322
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.