Learn More
Invitrogen™ SDCCAG3 Isoform 1 Polyclonal Antibody

Description
Recommended positive controls: Molt-4, mouse brain. Predicted reactivity: Mouse (90%), Rat (90%), Bovine (89%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | SDCCAG3 Isoform 1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 |
| Gene | ENTR1 |
| Gene Accession No. | A2AIW0, Q96C92 |
| Gene Alias | Antigen NY-CO-3; C330016H24Rik; C630038K21Rik; Endosome-associated-trafficking regulator 1; Entr1; NY-CO-3; RP11-413M3.3; Sdccag3; Serologically defined colon cancer antigen 3; serologically defined colon cancer antigen 3 homolog |
| Gene Symbols | ENTR1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.