Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SDCCAG8 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | SDCCAG8 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15669920
![]() |
Novus Biologicals
NBP15669920UL |
20 μL |
Each for $206.00
|
|
|||||
NBP156699
![]() |
Novus Biologicals
NBP156699 |
100 μL |
Each for $487.50
|
|
|||||
Description
SDCCAG8 Polyclonal specifically detects SDCCAG8 in Human samples. It is validated for Western Blot.Specifications
SDCCAG8 | |
Polyclonal | |
Rabbit | |
Q86SQ7 | |
10806 | |
Synthetic peptides corresponding to SDCCAG8(serologically defined colon cancer antigen 8) The peptide sequence was selected from the middle region of SDCCAG8. Peptide sequence IQCDQLRKELERQAERLEKELASQQEKRAIEKDMMKKEITKEREYMGSKM. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
CCCAPAntigen NY-CO-8, Centrosomal colon cancer autoantigen protein, hCCCAP, NPHP10, NY-CO-8, serologically defined colon cancer antigen 8, SLSN7 | |
SDCCAG8 | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title