Learn More
Invitrogen™ SDCCAG8 Polyclonal Antibody

Description
Immunogen sequence: QLELEKLKLT YEEKCEIEES QLKFLRNDLA EYQRTCEDLK EQLKHKEFLL AANTCNRVGG LCLKCAQHEA VLSQTHTNVH MQTIERLVKE RDDLMSAL Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 76%.
Specifications
Specifications
| Antigen | SDCCAG8 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | SDCCAG8 |
| Gene Accession No. | Q86SQ7 |
| Gene Alias | 2700048G21Rik; 5730470G24Rik; Antigen NY-CO-8; BBS16; CCCAP; CCCAP SLSN7; centrosomal colon cancer autoantigen protein; hCCCAP; HSPC085; mCCCAP; NPHP10; NY-CO-8; SDCCAG8; Serologically defined colon cancer antigen 8; serologically defined colon cancer antigen 8 homolog; SHH signaling and ciliogenesis regulator sdccag8; si:dkey-60b15.1; slinky; SLSN7 |
| Gene Symbols | SDCCAG8 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.