Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SDE2 Antibody, DyLight 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP119139G
Description
SDE2 Polyclonal antibody specifically detects SDE2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
SDE2 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
C1orf55, chromosome 1 open reading frame 55, dJ671D7.1, FLJ35382, hypothetical protein LOC163859, RP4-671D7.1, SDE2 telomere maintenance homolog (S. pombe) | |
The immunogen recognized by this antibody maps to a region between residue 350 and 400 of human chromosome 1 open reading frame 55 using the numbering given in entry NP_689821.3 (GeneID 163859). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 488 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
163859 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction