Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SDE2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556690
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556690 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556690 Supplier Invitrogen™ Supplier No. PA556690
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ETQEKKAESK EPIEEEPTGA GLNKDKETEE RTDGERVAEV APEERENVAV AKLQESQPGN AVIDKETIDL LAFTSVAELE LLGLEKLKCE LMA Highest antigen sequence identity to the following orthologs: Mouse - 37%, Rat - 40%.

Involved in both DNA replication and cell cycle control (PubMed:27906959). Unprocessed SDE2 interacts with PCNA via its PIP-box. The interaction with PCNA prevents monoubiquitination of the latter thereby inhibiting translesion DNA synthesis. The binding of SDE2 to PCNA also leads to processing of SDE2 by an unidentified deubiquitinating enzyme, cleaving off the N-terminal ubiquitin-like domain. The resulting mature SDE2 is degraded by the DCX(DTL) complex in a cell cycle- and DNA damage dependent manner (PubMed:27906959). Binding of SDE2 to PCNA is necessary to counteract damage due to ultraviolet light induced replication stress. The complete degradation of SDE2 is necessary to allow S-phase progression (PubMed:27906959). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen SDE2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 1.9 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SDE2
Gene Accession No. Q6IQ49
Gene Alias C1orf55; dJ671D7.1; FLJ35382; protein SDE2 homolog; Replication stress response regulator SDE2; RGD1305572; SDE2; SDE2 telomere maintenance homolog; SDE2 telomere maintenance homolog (S. pombe); UPF0667 protein C1orf55; UPF0667 protein C1orf55 homolog
Gene Symbols SDE2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SDE2. Recombinant protein control fragment (Product #RP-94304).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 163859
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.