Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SDE2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556691
Description
Immunogen sequence: MEGLETAEGS NSESSDDDSE EAPSTSGMGF HAPKIGSNGV EMAAKFPSGS QRARVVNTDH GSPEQLQIPV TDSGRHILED SCAELGESKE HMESRMVTET E Highest antigen sequence identity to the following orthologs: Mouse - 47%, Rat - 48%.
Involved in both DNA replication and cell cycle control (PubMed:27906959). Unprocessed SDE2 interacts with PCNA via its PIP-box. The interaction with PCNA prevents monoubiquitination of the latter thereby inhibiting translesion DNA synthesis. The binding of SDE2 to PCNA also leads to processing of SDE2 by an unidentified deubiquitinating enzyme, cleaving off the N-terminal ubiquitin-like domain. The resulting mature SDE2 is degraded by the DCX(DTL) complex in a cell cycle- and DNA damage dependent manner (PubMed:27906959). Binding of SDE2 to PCNA is necessary to counteract damage due to ultraviolet light induced replication stress. The complete degradation of SDE2 is necessary to allow S-phase progression (PubMed:27906959). [UniProt].
Specifications
SDE2 | |
Polyclonal | |
Unconjugated | |
SDE2 | |
C1orf55; dJ671D7.1; FLJ35382; protein SDE2 homolog; Replication stress response regulator SDE2; RGD1305572; SDE2; SDE2 telomere maintenance homolog; SDE2 telomere maintenance homolog (S. pombe); UPF0667 protein C1orf55; UPF0667 protein C1orf55 homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
163859 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.6 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6IQ49 | |
SDE2 | |
Recombinant protein corresponding to Human SDE2. Recombinant protein control fragment (Product #RP-94306). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction