Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SDF2 Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299669H
Description
SDF2 Polyclonal specifically detects SDF2 in Human samples. It is validated for ELISA.Specifications
SDF2 | |
Polyclonal | |
PBS | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
HRP | |
SDF-2, stromal cell-derived factor 2 | |
Produced in rabbits immunized with purified, recombinant SDF2 (NP_008854.2; Met1-Leu211). | |
0.1 mL | |
Signal Transduction | |
6388 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction