Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SDF2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583630
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583630 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583630 Supplier Invitrogen™ Supplier No. PA583630
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TVLCNGPYWV RDGEVRFKHS STEVLLSVTG E Highest antigen sequence indentity to the following orthologs: Mouse - 97%, Rat - 97%.

SDF2 is believed to be a secretory protein. It has regions of similarity to hydrophilic segments of yeast mannosyltransferases. Its expression is ubiquitous and the gene appears to be relatively conserved among mammals.The protein encoded by this gene is believed to be a secretory protein. It has regions of similarity to hydrophilic segments of yeast mannosyltransferases. Its expression is ubiquitous and the gene appears to be relatively conserved among mammals.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SDF2
Applications Immunocytochemistry
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SDF2
Gene Accession No. Q99470
Gene Alias AI853825; fk23f01; hypothetical protein LOC508463; Sdf2; SDF-2; stromal cell derived factor 2; stromal cell-derived factor 2; stromal cell-derived factor 2-like protein; wu:fk23f01; zgc:66291
Gene Symbols SDF2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SDF2. Recombinant protein control fragment (Product #RP-100141).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6388
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.