Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SDF2L1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552171
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552171 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552171 Supplier Thermo Scientific Supplier No. PA552171
Add to Cart
Add to Cart

SDF2L1 Polyclonal Antibody

SDF2L1 Polyclonal Antibody for IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen SDF2L1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SDF2L1
Gene Accession No. Q9HCN8
Gene Alias dihydropyrimidinase-like 2; OTTHUMT00000075032; PWP1-interacting protein 8; Sdf2l1; SDF2-like 1; SDF2-like protein 1; stromal cell derived factor 2 like 1; stromal cell-derived factor 2-like 1; stromal cell-derived factor 2-like protein 1; UNQ1941/PRO4424
Gene Symbols SDF2L1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SDF2L1. Recombinant protein control fragment (Product #RP-91344).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23753
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.