Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SDF2L1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA571817
Description
SDF2L1 Polyclonal Antibody for Western Blot, IHC (P), Flow
The specific function of the protein remains unknown.
Specifications
SDF2L1 | |
Polyclonal | |
Unconjugated | |
SDF2L1 | |
dihydropyrimidinase-like 2; OTTHUMT00000075032; PWP1-interacting protein 8; Sdf2l1; SDF2-like 1; SDF2-like protein 1; stromal cell derived factor 2 like 1; stromal cell-derived factor 2-like 1; stromal cell-derived factor 2-like protein 1; UNQ1941/PRO4424 | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
23753, 64136 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
0.4 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q9ESP1, Q9HCN8 | |
SDF2L1 | |
KLH conjugated synthetic peptide between 160-189 amino acids from the C-terminal region of human SDF2L1. | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction