Learn More
Invitrogen™ SDF4 Polyclonal Antibody

Description
SDF4 Polyclonal Antibody for Western Blot, ELISA
Specifications
Specifications
| Antigen | SDF4 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 |
| Gene | SDF4 |
| Gene Accession No. | Q9BRK5 |
| Gene Alias | 45 kDa calcium-binding protein; cab45; PSEC0034; Sdf4; SDF-4; stromal cell derived factor 4; stromal cell-derived factor 4 |
| Gene Symbols | SDF4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.