Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SDF4 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA5120363
Encompass
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120363 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120363 Supplier Thermo Scientific Supplier No. PA5120363
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: Mouse brain, Mouse kidney, Mosue liver, Mouse pancreas, Rat heart. The target is usually found in the following locations: Cell projection, Cytoplasm, Golgi apparatus lumen, bleb. Immunogen sequence: RPANHSSTRE RVANREENEI LPPDHLNGVK LEMDGHLNRG FHQEVFLGKD LGGFDEDAEP RRSRRKLMVI FSKVDVNTDR KISAKEMQRW IMEKTAEHFQ EAMEESKTHF RAVDPDGDGH VSWDEYKVKF LASKGHSEKE VADAIRLNEE LKVDEETQEV LENL

This gene encodes a stromal cell derived factor that is a member of the CREC protein family. The encoded protein contains six EF-hand motifs and calcium-binding motifs. This protein localizes to the Golgi lumen and may be involved in regulating calcium dependent cellular activities.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SDF4
Applications Western Blot
Classification Polyclonal
Concentration 2.18 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene SDF4
Gene Accession No. Q61112, Q91ZS3
Gene Alias 45 kDa calcium-binding protein; cab45; PSEC0034; Sdf4; SDF-4; stromal cell derived factor 4; stromal cell-derived factor 4
Gene Symbols SDF4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 37-200 of human SDF4 (NP_057260.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 155173, 20318
Target Species Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.