Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SDFR1/NPTN Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984164
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4164 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4164 Supplier Bioss Supplier No. BS1801R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a type I transmembrane protein belonging to the Ig superfamily. The protein is believed to be involved in cell-cell interactions or cell-substrate interactions. Alternative splicing results in multiple transcript variants.

Specifications

Antigen SDFR1/NPTN
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene NPTN
Gene Accession No. Q9Y639
Gene Alias AW554172; glycoprotein 55; Glycoprotein 55/65; glycoprotein 65; GP55; gp55/65; GP65; Neuroplastin; np55; np65; NPTN; SDFR1; Sdr1; SDR-1; stromal cell derived factor receptor 1; Stromal cell-derived receptor 1
Gene Symbols NPTN
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human NPTN.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27020
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.