Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SDHA Monoclonal Antibody (GT20710)
GREENER_CHOICE

Catalog No. PIMA527782
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527782 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527782 Supplier Invitrogen™ Supplier No. MA527782
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive Control: 293T Predicted Reactivity: Mouse (98%), Zebrafish (97%), Drosophila (87%), Xenopus laevis (82%), Chicken (97%), Bovine (99%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a major catalytic subunit of succinate-ubiquinone oxidoreductase, a complex of the mitochondrial respiratory chain. The complex is composed of four nuclear-encoded subunits and is localized in the mitochondrial inner membrane. Mutations in this gene have been associated with a form of mitochondrial respiratory chain deficiency known as Leigh Syndrome. A pseudogene has been identified on chromosome 3q29.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SDHA
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone GT20710
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and no preservative; pH 7
Gene SDHA
Gene Accession No. P31040, Q8K2B3, Q920L2
Gene Alias 1500032O14Rik; 2310034D06Rik; 4921513A11; C81073; CMD1GG; Flavoprotein subunit of complex II; FP; im:7141001; mitochondrial succinate dehydrogenase complex subunit A; PGL5; SDH1; SDH2; SDHA; SDHA protein; SDHF; SDHFP1; si:dkeyp-84f11.2; succinate dehydrogenase [ubiquinone] flavoprotein subunit; succinate dehydrogenase [ubiquinone] flavoprotein subunit, mitochondrial; succinate dehydrogenase complex flavoprotein subunit A; succinate dehydrogenase complex subunit A; succinate dehydrogenase complex subunit A flavoprotein; succinate dehydrogenase complex subunit A, flavoprotein (Fp); succinate dehydrogenase complex, subunit A, flavoprotein (Fp); succinate dehydrogenase flavoprotein subunit; succinate dehydrogenase flavoprotein subunit A; zgc:56051
Gene Symbols SDHA
Host Species Mouse
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human SDHA. The exact sequence is proprietary.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 157074, 6389, 66945
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.