Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SDHA Rabbit anti-Human, Alexa Fluor 405, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299321AF405
Description
SDHA Polyclonal specifically detects SDHA in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
SDHA | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Alexa Fluor 405 | |
CMD1GG, EC 1.3.5.1, Fp, SDH1, SDH2, SDHFmitochondrial, succinate dehydrogenase complex flavoprotein subunit, succinate dehydrogenase complex, subunit A, flavoprotein (Fp) | |
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human SDHA. | |
0.1 mL | |
Hypoxia | |
6389 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction