Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SDHAF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA519194
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA519194 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA519194 Supplier Invitrogen™ Supplier No. PA519194
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with canine and mouse based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.

SDHAF1 (Succinate dehydrogenase complex assembly factor 1) is one of the subunits of the succinate dehydrogenase (SDH) complex (or complex II) of the mitochondrial respiratory chain. SDHAF1 resides in the mitochondria, and is essential for SDH assembly, but does not physically associate with the complex in vivo. Mutations in this gene are associated with SDH-defective infantile leukoencephalopathy (mitochondrial complex II deficiency). Unlike the related protein SDHAF2, SDHAF1 is not thought to be a tumor suppressor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SDHAF1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene SDHAF1
Gene Accession No. A6NFY7
Gene Alias 0610010E21Rik; AI430885; AW490662; LYR motif containing 8; LYR motif-containing protein 8; LYRM8; SDH assembly factor 1; Sdhaf1; Succinate dehydrogenase assembly factor 1, mitochondrial; succinate dehydrogenase complex assembly factor 1
Gene Symbols SDHAF1
Host Species Goat
Immunogen Synthetic peptide sequence (HDSTGAPETRPDGR) corresponding to the C-terminus amino acids of SDHAF1 (aa 102-115).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 644096
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.