Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SDHAF3 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA5140050

Catalog No. PIPA5140050


Only null left
Add to Cart
This item is not returnable. View return policy

Description

Description

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: GRHVSRVRAL YKRVLQLHRV LPPDLKSLGD QYVKDEFRRH KTVGSDEAQR FLQEWEAR.

Plays an essential role in the assembly of succinate dehydrogenase (SDH), an enzyme complex (also referred to as respiratory complex II) that is a component of both the tricarboxylic acid (TCA) cycle and the mitochondrial electron transport chain, and which couples the oxidation of succinate to fumarate with the reduction of ubiquinone (coenzyme Q) to ubiquinol. Promotes maturation of the iron-sulfur protein subunit SDHB of the SDH catalytic dimer, protecting it from the deleterious effects of oxidants. May act together with SDHAF1.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SDHAF3
Polyclonal
Unconjugated
SDHAF3
0610005M07Rik; 4933430A16Rik; Acn9; ACN9 homolog; ACN9 homolog (S. cerevisiae); DC11; Liver regeneration-related protein LRRGT00092; LRRGT00092; LYRM10; protein ACN9 homolog, mitochondrial; SDH assembly factor 3; Sdh7; SDHAF3; Succinate dehydrogenase assembly factor 3, mitochondrial; succinate dehydrogenase complex assembly factor 3
Rabbit
Antigen affinity chromatography
RUO
57001
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9NRP4
SDHAF3
Recombinant protein corresponding to Human SDHAF3. Recombinant protein control fragment (Product #RP-108740).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.