Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SDHAF4 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$206.50 - $426.00

Specifications

Antigen C6orf57
Concentration 0.5 mg/mL
Dilution Western Blot 5-10ug/ml
Applications Western Blot
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP227170SS
SDP
View Documents
Novus Biologicals
NBP227170SS
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NBP227170
SDP
View Documents
Novus Biologicals
NBP227170
0.1 mg
Each for $426.00
Only null left
Add to Cart
 
Description

Description

SDHAF4 Polyclonal specifically detects SDHAF4 in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

C6orf57
Western Blot 5-10ug/ml
Polyclonal
Purified
RUO
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
chromosome 6 open reading frame 57, hypothetical protein LOC135154, MGC104225
C6ORF57
IgG
Protein A purified
The amino acid sequence used as immunogen is 100% homologous in mouse, and 94% homologous in human, monkey, dog and rat.
0.5 mg/mL
Western Blot
Unconjugated
Rabbit
Mouse
Q8BTE0
135154
A portion of amino acids 75-129 of mouse UPF0369 protein C6orf57 homolog was used as the immunogen.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.