Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SDHB Antibody, Alexa Fluor™ 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP154154AF405
Description
SDHB Polyclonal specifically detects SDHB in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
SDHB | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
P21912 | |
SDHB | |
A genomic peptide made to an N-terminal region of the human SDHB protein (within residues 1-150). [Swiss-Prot P21912] | |
0.1 mL | |
Cancer | |
6390 | |
Human, Mouse | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
EC 1.3.5.1, FLJ92337, IP, Iron-sulfur subunit of complex II, PGL4, SDH, SDH1, SDH2, SDHIP, succinate dehydrogenase [ubiquinone] iron-sulfur subunit, mitochondrial, succinate dehydrogenase complex, subunit B, iron sulfur (Ip) | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human and mouse. Immunogen sequence has 92% identity to rat, cow and pig, 85% identity to Xenopus and 84% identity to Zebrafish. | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction