Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SDHB Antibody (SDHB/2382), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308537V
Description
SDHB Monoclonal antibody specifically detects SDHB in Human samples. It is validated for Western Blot, Protein ArraySpecifications
SDHB | |
Monoclonal | |
DyLight 405 | |
EC 1.3.5.1, FLJ92337, IP, Iron-sulfur subunit of complex II, PGL4, SDH, SDH1, SDH2, SDHIP, succinate dehydrogenase [ubiquinone] iron-sulfur subunit, mitochondrial, succinate dehydrogenase complex, subunit B, iron sulfur (Ip) | |
Recombinant fragment (around aa 165-273) of human SDHB protein (exact sequence is proprietary) (Uniprot: P21912) | |
100 μg | |
Cancer | |
6390 | |
Store at 4°C in the dark. | |
IgG2b κ |
Western Blot, Protein Array | |
SDHB/2382 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction