Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SDHB Recombinant Rabbit Monoclonal Antibody (9S1A0)
GREENER_CHOICE

Catalog No. PIMA535191
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA535191 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA535191 Supplier Invitrogen™ Supplier No. MA535191
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: DGLYECILCA CCSTSCPSYW WNGDKYLGPA VLMQAYRWMI DSRDDFTEER LAKLQDPFSL YRCHTIMNCT RTCPKGLNPG KAIAEIKKMM ATYKEKKASV.

Complex II of the respiratory chain, which is specifically involved in the oxidation of succinate, carries electrons from FADH to CoQ. The complex is composed of four nuclear-encoded subunits and is localized in the mitochondrial inner membrane. The iron-sulfur subunit is highly conserved and contains three cysteine-rich clusters which may comprise the iron-sulfur centers of the enzyme. Sporadic and familial mutations in this gene result in paragangliomas and pheochromocytoma, and support a link between mitochondrial dysfunction and tumorigenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SDHB
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 9S1A0
Concentration 0.35 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Sdhb
Gene Accession No. P21912, P21913, Q9CQA3
Gene Alias 0710008N11Rik; CWS2; fj46d06; FLJ92337; I79_022285; Ip; iron-sulfur subunit; iron-sulfur subunit of complex II; OTTHUMP00000044624; PGL4; SDH; SDH1; SDH2; Sdhb; SDHB protein; SDHB; sdhB; SDHIP; succinate dehydrogenase [ubiquinone] iron-sulfur subunit; Succinate dehydrogenase [ubiquinone] iron-sulfur subunit, mitochondrial; succinate dehydrogenase complex iron sulfur subunit B; succinate dehydrogenase complex subunit B; succinate dehydrogenase complex subunit B, iron sulfur (Ip); succinate dehydrogenase complex, subunit B, iron sulfur (Ip); succinate dehydrogenase Ip subunit; succinate dehydrogenase iron-protein subunit; wu:fj46d06
Gene Symbols Sdhb
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 181-280 of human SDHB (P21912).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 298596, 6390, 67680
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.