Learn More
SDHB Mouse anti-Human, Clone: OTI2A3, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF808201

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Complex II of the respiratory chain, which is specifically involved in the oxidation of succinate, carries electrons from FADH to CoQ. The complex is composed of four nuclear-encoded subunits and is localized in the mitochondrial inner membrane. The iron-sulfur subunit is highly conserved and contains three cysteine-rich clusters which may comprise the iron-sulfur centers of the enzyme. Sporadic and familial mutations in this gene result in paragangliomas and pheochromocytoma, and support a link between mitochondrial dysfunction and tumorigenesis.Specifications
SDHB | |
Monoclonal | |
Unconjugated | |
SDHB | |
CWS2, IP, PGL4, SDH, SDH1, SDH2, SDHIP | |
Mouse | |
Affinity Chromatography | |
RUO | |
6390 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI2A3 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P21912 | |
SDHB | |
Full length human recombit protein of human SDHB produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.