Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SDHB Rabbit anti-Human, Unconjugated, Clone: ABDF-19, Abnova™
Rabbit Monoclonal Antibody.
Supplier: Abnova Corporation MAB20699
Description
SDHB Monoclonal antibody specifically detects SDHB in Human samples. It is validated for IP, IHC, WB-Ce, Flow Cyt.Specifications
SDHB | |
Monoclonal | |
Unconjugated | |
SDHB | |
Rabbit | |
100 μg | |
6390 | |
Store at -20°C for one year. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing. | |
IgG |
Immunoprecipitation, Immunohistochemistry, Western Blot, Flow Cytometry | |
ABDF-19 | |
succinate dehydrogenase complex, subunit B, iron sulfur (Ip) | |
FLJ92337, IP, PGL4, SDH, SDH1, SDHIP | |
A synthetic peptide corresponding to human SDHB | |
Primary | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction