Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SDHC Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114213
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114213 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114213 Supplier Invitrogen™ Supplier No. PA5114213
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YSWSLPMAMS ICHRGTGIAL SADVGPRKRP EDSPAIPVWS GCPGSYCVVL For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Human: 100%.

This gene encodes one of four nuclear-encoded subunits that comprise succinate dehydrogenase, also known as mitochondrial complex II, a key enzyme complex of the tricarboxylic acid cycle and aerobic respiratory chains of mitochondria. The encoded protein is one of two integral membrane proteins that anchor other subunits of the complex, which form the catalytic core, to the inner mitochondrial membrane. There are several related pseudogenes for this gene on different chromosomes. Mutations in this gene have been associated with paragangliomas. Alternatively spliced transcript variants have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SDHC
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SDHC
Gene Accession No. Q99643
Gene Alias 0610010E03Rik; AI316496; AU019277; CYB560; CYBL; cytochrome B large subunit of complex II; integral membrane protein CII-3; integral membrane protein CII-3b; large subunit of cytochrome b; PGL3; QPs1; QPs-1; SDH3; SDHC; succinate dehydrgenase cytochrome b; succinate dehydrogenase 3, integral membrane subunit; succinate dehydrogenase complex subunit C; succinate dehydrogenase complex subunit C integral membrane protein 15kDa; succinate dehydrogenase complex, subunit C, integral membrane protein; succinate dehydrogenase complex, subunit C, integral membrane protein, 15kD; succinate dehydrogenase complex, subunit C, integral membrane protein, 15kDa; succinate dehydrogenase cytochrome b560 subunit, mitochondrial; Succinate-ubiquinone oxidoreductase cytochrome B large subunit
Gene Symbols SDHC
Host Species Rabbit
Immunogen Immunogen sequence: YSWSLPMAMS ICHRGTGIAL SADVGPRKRP EDSPAIPVWS GCPGSYCVVL.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6391
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.