Learn More
SDHD Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | SDHD |
---|---|
Concentration | 167 μg/mL |
Applications | Immunofluorescence, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-223-8522
![]() ![]() |
Proteintech Group Inc
265191AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The SDHD antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human SDHD. This antibody recognizes human antigen. The SDHD antibody has been validated for the following applications: IF, ELISA analysis.
Specifications
SDHD | |
Immunofluorescence, ELISA | |
Unconjugated | |
Human | |
IgG |
167 μg/mL | |
Polyclonal | |
Rabbit | |
6392 | |
Primary |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.