Learn More
Spectrum Solutions™ SDNA-1000 Molecular Diagnostic Saliva Collection Device
Catalog No. 22140000
Change view
Click to view available options
Quantity:
360/Cs.
Catalog No. Quantity
22-140-000 360/Cs.
1 options
This item is not returnable. View return policy
Catalog No. 22-140-000 Supplier Spectrum Solutions™ Supplier No. SDNA1000
Add to Cart
Add to Cart
This item is not returnable. View return policy
Engineered to lead the saliva collection market in molecular diagnostic applications. This self-contained, EUA authorized saliva collection system provides sample consistency while suspending and neutralizing viral RNA transcripts post-collection for sensitive and specific analysis.
  • Creates the most robust and safest biomaterial collection approach for the detection of COVID-19 infections
  • Leads the way to a new era of at-home biosample self-collection for the diagnosis of viral infections
  • First EUA authorized saliva collection kit for COVID-19 testing
  • First EUA authorized device for direct-to-patient at-home saliva sample self-collection
  • Intuitive, 3-step design is proven to reduce customer collection errors
  • Single device for both DNA & viral RNA applications
  • 100% inactivation of the live virus at ambient temps with EUA validated viral inactivation data
  • Delivers the safest & most robust biomaterial for detecting COVID-19 
  • Provides over 10 days of post-collection stability with no degradation in sample efficacy
  • Collection system maintainence of biosample consistency significantly reduces costs associated with sample failures & recollection
  • Mitigates any risk of infection throughout the testing process 
  • Pinpoints life-cycle stage of active viral infection
  • In device viral neutralization eliminates UN3373 designation delivering UPS and Fedex shipping clearance
  • Patented preservation chemistry enables easy extraction & purification for sensitivity boost & optimal accuracy in molecular analysis
  • Tube compatible with automated platforms & customizable for incremental efficiency improvements
  • Qualified commercial RNA extraction chemistries include Perkin Elmer, Thermo Fisher, Roche, Qiagen, and more
Specifications
Certifications/Compliance FDA, EUA Device Certification (EUA202432)
Content And Storage Sealed Blister Pack with Spectrum Tyvek Label, Spit Tube with Attached Funnel, Spectrum’s Patented Blue Preservation Chemistry Filled Cap, Device Instructions for use (IFU) in 3 Languages, and Unique Barcode Serialization for Identification and Sample Chain-of-Custody
For Use With (Application) COVID-19 Diagnostic Testing, Clinical Research, Molecular Diagnostic Applications, Personalized Skincare, Health & Wellness, Direct-to-Consumer Biosample Collection, and Biobanking
Quantity 360/Cs.