Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SDPR Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA534521
Description
A suggested positive control is Hela cell lysate. PA5-34521 can be used with blocking peptide PEP-1564.
The serum deprivation-response protein (SDPR) is a calcium-independent phospholipid-binding protein whose expression is increased in serum-starved cells. SDPR is a substrate for protein kinase C (PKC) phosphorylation and recruits the polymerase I and transcript release factor (PTRF) to caveolae. Removal of this protein causes caveolae loss and its over-expression results in caveolae deformation and membrane tubulation. Both SDPR and PTRF, as well as the other member of the cavin family PRKCDBP were down regulated in breast cancer cell lines and breast tumor tissue, suggesting that expression of the cavin family proteins could be a useful prognostic indicator of breast cancer progression.
Specifications
SDPR | |
Polyclonal | |
Unconjugated | |
CAVIN2 | |
caveolae associated protein 2; caveolae-associated protein 2; cavin 2; CAVIN2; cavin-2; EMBL:AAH81956.1}; phosphatidylserine binding protein; phosphatidylserine-binding protein; PS-p68; SDPR; sdpr {ECO:0000312; SDR; serum deprivation response; serum deprivation response protein; serum deprivation-response protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8436 | |
Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
O95810 | |
CAVIN2 | |
An 18 amino acid peptide near the amino terminus of human SDPR. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction